The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008842.3 | NP_032868.2 | 5 - 6 | 890 | 67 |
GenBank mRNA | AK154127.1 | - | - | 233 | 67 |
AK154669.1 | - | 4 - 5 | 729 | 67 | |
BC042885.1 | - | 5 - 6 | 1123 | 67 | |
BC053019.1 | - | 5 - 6 | 890 | 67 | |
BC055316.1 | - | 5 - 6 | 890 | 67 |