The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252565.1 | NP_001239494.1 | 2 - 3 | 396 | 92 |
NM_011012.5 | NP_035142.1 | 2 - 3 | 468 | 92 | |
GenBank mRNA | AF062381.1 | - | 1 - 2 | 242 | 92 |
AF075605.1 | - | 1 - 2 | 241 | 92 | |
AK038620.1 | - | 2 - 3 | 398 | 92 | |
AK079529.1 | - | 2 - 3 | 441 | 92 | |
BC050885.1 | - | 2 - 3 | 390 | 92 | |
BC051982.1 | - | 2 - 3 | 328 | 92 | |
U04952.1 | - | 2 - 3 | 389 | 92 | |
U09421.1 | - | 2 - 3 | 519 | 92 | |
U14165.1 | - | 1 - 2 | 229 | 92 | |
X91813.1 | - | 2 - 3 | 399 | 92 |