The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038663.1 | NP_001033752.1 | 1 - 2 | 358 | 117 |
NM_011949.3 | NP_036079.1 | 1 - 2 | 358 | 117 | |
GenBank mRNA | AK035386.1 | - | 1 - 2 | 332 | 117 |
AK048127.1 | - | 1 - 2 | 360 | 117 | |
AK087925.1 | - | 1 - 2 | 333 | 117 | |
AK132241.1 | - | 1 - 2 | 346 | 117 | |
AK148853.1 | - | 1 - 2 | 338 | 117 | |
AK205643.1 | - | 1 - 2 | 52 | 117 | |
AK216848.1 | - | 1 - 2 | 60 | 117 | |
BB648734.1 | - | 1 - 2 | 359 | 117 | |
BC058258.1 | - | 1 - 2 | 301 | 117 | |
D10939.1 | - | 1 - 2 | 173 | 117 | |
X58712.1 | - | 1 - 2 | 131 | 117 |