Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a8
Entrez Gene ID
56017
Gene Name
solute carrier family 2, (facilitated glucose transporter), member 8
Gene Aliases
D2Ertd44e, GLUT8, GlutX1
Chromosome Location
Chr.2: 32972981 - 32982113 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019488.4 NP_062361.1 1 - 2 103 72
GenBank mRNA AJ245936.1 - 1 - 2 80 72
AK081806.1 - 1 - 2 120 72
AK141193.1 - 1 - 2 116 72
AK155792.1 - 1 - 2 131 72
AK170319.1 - 1 - 2 116 72
AY856043.1 - 1 - 2 64 72
AY856044.1 - 1 - 2 64 72
AY856045.1 - 1 - 2 64 72
BC090993.1 - 1 - 2 103 72
Y17802.1 - 1 - 2 85 72

View Full Product Details