Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a8
Entrez Gene ID
56017
Gene Name
solute carrier family 2, (facilitated glucose transporter), member 8
Gene Aliases
D2Ertd44e, GLUT8, GlutX1
Chromosome Location
Chr.2: 32972981 - 32982113 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019488.4 NP_062361.1 5 - 6 768 110
GenBank mRNA AF232061.2 - 5 - 6 725 110
AJ245936.1 - 5 - 6 745 110
AK081806.1 - 5 - 6 785 110
AK141193.1 - 4 - 5 934 110
AK170319.1 - 5 - 6 781 110
AY856043.1 - 5 - 6 729 110
AY856044.1 - 5 - 6 729 110
AY856045.1 - 5 - 6 729 110
AY856046.1 - 2 - 3 259 110
BC021758.1 - 1 - 2 94 110
BC090993.1 - 5 - 6 768 110
Y17802.1 - 5 - 6 750 110

View Full Product Details