Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kng1
Entrez Gene ID
16644
Gene Name
kininogen 1
Gene Aliases
Kng
Chromosome Location
Chr.16: 23058220 - 23082078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102411.1 NP_001095881.1 4 - 5 712 130
NM_001102412.1 NP_001095882.1 4 - 5 712 130
NM_023125.3 NP_075614.1 4 - 5 712 130
GenBank mRNA AK005547.1 - 4 - 5 677 130
AY462058.1 - 4 - 5 592 130
AY660571.1 - 4 - 5 619 130
BC018158.1 - 4 - 5 664 130
BC108936.1 - 4 - 5 668 130
D84415.1 - 4 - 5 592 130
D84435.1 - 4 - 5 592 130

View Full Product Details