Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cck
Entrez Gene ID
12424
Gene Name
cholecystokinin
Gene Aliases
-
Chromosome Location
Chr.9: 121489824 - 121495694 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284508.1 NP_001271437.1 1 - 2 92 56
NM_031161.4 NP_112438.1 1 - 2 92 56
GenBank mRNA AK002677.1 - 1 - 2 60 56
BC028487.1 - 1 - 2 37 56
BY127153.1 - 1 - 2 61 56

View Full Product Details