The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016694.3 | NP_057903.1 | 11 - 12 | 1379 | 59 |
GenBank mRNA | AB019558.1 | - | 11 - 12 | 1350 | 59 |
AF250293.1 | - | 11 - 12 | 1379 | 59 | |
AK015990.1 | - | 2 - 3 | 363 | 59 | |
BC113204.1 | - | 11 - 12 | 1362 | 59 | |
FJ555554.1 | - | 9 - 10 | 1132 | 59 | |
FJ555555.1 | - | 6 - 7 | 790 | 59 | |
FJ555556.1 | - | 10 - 11 | 1112 | 59 | |
FJ555557.1 | - | 10 - 11 | 1237 | 59 | |
GU272056.1 | - | 5 - 6 | 653 | 59 |