Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pdcd1lg2
- Entrez Gene ID
-
58205
- Gene Name
- programmed cell death 1 ligand 2
- Gene Aliases
-
B7-DC, Btdc, F730015O22Rik, PD-L2
- Chromosome Location
-
Chr.19: 29406240 - 29472927 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF142780.2 |
- |
5 - 6 |
975 |
117 |
View Full Product Details