The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021433.3 | NP_067408.1 | 6 - 7 | 788 | 142 |
GenBank mRNA | AF237814.1 | - | 6 - 7 | 598 | 142 |
AK009690.1 | - | 6 - 7 | 743 | 142 | |
AK018167.1 | - | 6 - 7 | 142 | ||
AK019106.1 | - | 6 - 7 | 750 | 142 | |
AK031673.1 | - | 6 - 7 | 767 | 142 | |
AK044905.1 | - | 6 - 7 | 749 | 142 | |
AK049124.1 | - | 6 - 7 | 748 | 142 | |
AK151549.1 | - | 6 - 7 | 742 | 142 | |
AK163167.1 | - | - | 161 | 142 | |
BC029205.1 | - | 6 - 7 | 705 | 142 | |
CN456515.1 | - | 2 - 3 | 168 | 142 |