Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mgat5
Entrez Gene ID
107895
Gene Name
mannoside acetylglucosaminyltransferase 5
Gene Aliases
4930471A21Rik, 5330407H02Rik, AI480971, GlcNAc-TV
Chromosome Location
Chr.1: 127204977 - 127488334 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145128.3 NP_660110.2 11 - 12 1877 61
GenBank mRNA AF474154.1 - 9 - 10 1257 61
AF474155.1 - 9 - 10 1320 61
AK030402.1 - 11 - 12 1845 61
BC125533.1 - 9 - 10 1285 61
BC125535.1 - 9 - 10 1285 61

View Full Product Details