Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St8sia3
Entrez Gene ID
20451
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
AI847333, ST8SiaIII, Siat8c
Chromosome Location
Chr.18: 64254359 - 64276144 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009182.3 NP_033208.2 3 - 4 972 75
GenBank mRNA AK015874.1 - 3 - 4 956 75
AK133780.1 - 1 - 2 257 75
AK134678.1 - 1 - 2 268 75
AK161482.1 - 3 - 4 973 75
BC075645.1 - 1 - 2 267 75
X80502.1 - 1 - 2 257 75

View Full Product Details