The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145215.2 | NP_660250.1 | 1 - 2 | 546 | 74 |
GenBank mRNA | AF412033.1 | - | 1 - 2 | 546 | 74 |
AK004244.1 | - | 1 - 2 | 81 | 74 | |
BC069866.1 | - | 1 - 2 | 168 | 74 |