The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153537.4 | NP_705765.3 | 20 - 21 | 3980 | 94 |
GenBank mRNA | AK052352.1 | - | 9 - 10 | 1340 | 94 |
AK122336.1 | - | 18 - 19 | 3899 | 94 | |
BC025856.1 | - | 2 - 3 | 178 | 94 | |
BC058712.1 | - | 20 - 21 | 3976 | 94 | |
BC069853.1 | - | 1 - 2 | 107 | 94 |