Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
244431
Gene Name
sarcoglycan zeta
Gene Aliases
C230085N17Rik
Chromosome Location
Chr.8: 37517840 - 38661508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145841.2 NP_665840.2 5 - 6 1290 99
GenBank mRNA AK048960.1 - 5 - 6 1254 99
AK136779.1 - 5 - 6 1291 99
AY095374.1 - 4 - 5 506 99
AY250713.1 - 4 - 5 506 99
BC104048.1 - 4 - 5 506 99
BC104049.2 - 4 - 5 506 99

View Full Product Details