Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn6
Entrez Gene ID
14976
Gene Name
prefoldin subunit 6
Gene Aliases
H-2Ke2, H2-Ke2, Ke-2
Chromosome Location
Chr.17: 33938909 - 33940343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185182.1 NP_001172111.1 2 - 3 110 71
NM_010385.2 NP_034515.1 1 - 2 314 71
GenBank mRNA AA023721.1 - 1 - 2 90 71
AK010679.1 - 1 - 2 304 71
AK076015.1 - 1 - 2 297 71
AK167900.1 - 1 - 2 300 71
AK177303.1 - 1 - 2 182 71
BC022974.1 - 1 - 2 280 71
BY238879.1 - 1 - 2 314 71
KJ650212.1 - 1 - 2 58 71
M65255.1 - 1 - 2 277 71

View Full Product Details