The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185182.1 | NP_001172111.1 | 4 - 5 | 305 | 69 |
NM_010385.2 | NP_034515.1 | 3 - 4 | 509 | 69 | |
GenBank mRNA | AK010679.1 | - | 3 - 4 | 499 | 69 |
AK076015.1 | - | 3 - 4 | 492 | 69 | |
AK167900.1 | - | 3 - 4 | 495 | 69 | |
AV446044.1 | - | 2 - 3 | 164 | 69 | |
BC022974.1 | - | 3 - 4 | 475 | 69 | |
KJ650212.1 | - | 3 - 4 | 253 | 69 |