The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113573.1 | NP_001107045.1 | 11 - 12 | 2253 | 103 |
NM_001113574.1 | NP_001107046.1 | 12 - 13 | 2304 | 103 | |
NM_023336.4 | NP_075825.3 | 11 - 12 | 2413 | 103 | |
GenBank mRNA | AB206708.2 | - | 11 - 12 | 2115 | 103 |
AF269193.1 | - | 11 - 12 | 2201 | 103 | |
AK009979.1 | - | 2 - 3 | 405 | 103 | |
AK010739.1 | - | 4 - 5 | 838 | 103 | |
AK014429.1 | - | 3 - 4 | 465 | 103 | |
AK136429.1 | - | 11 - 12 | 2185 | 103 | |
AK159827.1 | - | 11 - 12 | 2239 | 103 | |
AK160752.1 | - | 11 - 12 | 2212 | 103 | |
AK161164.1 | - | 10 - 11 | 3447 | 103 | |
AK167573.1 | - | 11 - 12 | 2211 | 103 | |
AY513269.1 | - | 10 - 11 | 2079 | 103 | |
BC031536.1 | - | 11 - 12 | 2227 | 103 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK142230.1 | - | Chr. - - [-] on Build | 103 |