The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024256.2 | NP_077218.1 | 1 - 2 | 257 | 108 |
GenBank mRNA | AB019523.1 | - | 1 - 2 | 136 | 108 |
AK048428.1 | - | 1 - 2 | 256 | 108 | |
AK140424.1 | - | 1 - 2 | 223 | 108 | |
AK144402.1 | - | 1 - 2 | 276 | 108 | |
AK146264.1 | - | 1 - 2 | 168 | 108 | |
BC002103.1 | - | 1 - 2 | 124 | 108 | |
BC004038.1 | - | 1 - 2 | 257 | 108 | |
BC012930.1 | - | 1 - 2 | 245 | 108 |