Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmd
Entrez Gene ID
67468
Gene Name
monocyte to macrophage differentiation-associated
Gene Aliases
1200017E07Rik, 1810073C06Rik, AA690185
Chromosome Location
Chr.11: 90187687 - 90278574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026178.2 NP_080454.1 6 - 7 752 85
GenBank mRNA AK004819.1 - 6 - 7 706 85
AK011418.1 - 6 - 7 717 85
AK149704.1 - 6 - 7 702 85
AK158577.1 - 6 - 7 710 85
AK171637.1 - 6 - 7 706 85
AY424300.1 - 6 - 7 592 85
BC021914.1 - 6 - 7 752 85
BC034117.1 - 7 - 8 1104 85

View Full Product Details