Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Odf3
Entrez Gene ID
69287
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
1700011O04Rik, SHIPPO1
Chromosome Location
Chr.7: 140847916 - 140850925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027019.3 NP_081295.2 6 - 7 702 118
GenBank mRNA AB067773.1 - 6 - 7 791 118
AK005883.1 - 6 - 7 813 118
BC104105.1 - 6 - 7 702 118
BC104106.1 - 6 - 7 702 118
BC106806.1 - 6 - 7 702 118

View Full Product Details