The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053093.1 | NP_444323.1 | 1 - 2 | 292 | 84 |
GenBank mRNA | AF235035.1 | - | 1 - 2 | 292 | 84 |
AF515827.1 | - | 1 - 2 | 126 | 84 | |
BC119426.1 | - | 1 - 2 | 272 | 84 |