Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Endou
Entrez Gene ID
19011
Gene Name
endonuclease, polyU-specific
Gene Aliases
Pp11r, Tcl-30
Chromosome Location
Chr.15: 97711015 - 97731405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168693.1 NP_001162164.1 9 - 10 1256 102
NM_008902.3 NP_032928.1 8 - 9 1255 102
GenBank mRNA AK132616.1 - 9 - 10 1197 102
AK169457.1 - 7 - 8 1346 102
M95545.1 - 8 - 9 1255 102

View Full Product Details