The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009694.3 | NP_033824.1 | 1 - 2 | 248 | 122 |
GenBank mRNA | AF161699.1 | - | 1 - 2 | 249 | 122 |
AK009047.1 | - | 1 - 2 | 146 | 122 | |
BC027530.1 | - | 1 - 2 | 158 | 122 |