The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012060.4 | NP_036190.2 | 5 - 6 | 580 | 111 |
GenBank mRNA | AK008043.1 | - | 5 - 6 | 580 | 111 |
AK011500.1 | - | 5 - 6 | 581 | 111 | |
AK151641.1 | - | 5 - 6 | 581 | 111 | |
BC002106.1 | - | 5 - 6 | 548 | 111 | |
X81816.1 | - | 4 - 5 | 480 | 111 |