The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013464.4 | NP_038492.1 | 7 - 8 | 1258 | 82 |
GenBank mRNA | AF405560.1 | - | 7 - 8 | 892 | 82 |
AF405561.1 | - | 7 - 8 | 892 | 82 | |
AF405562.1 | - | 7 - 8 | 892 | 82 | |
AF405563.1 | - | 7 - 8 | 892 | 82 | |
AF405565.1 | - | 7 - 8 | 892 | 82 | |
AF405566.1 | - | 7 - 8 | 892 | 82 | |
AF405567.1 | - | 7 - 8 | 892 | 82 | |
AF405568.1 | - | 7 - 8 | 892 | 82 | |
AF405570.1 | - | 7 - 8 | 892 | 82 | |
AK136072.1 | - | 1 - 2 | 142 | 82 | |
AK153678.1 | - | 7 - 8 | 1259 | 82 | |
AK155380.1 | - | 7 - 8 | 1310 | 82 | |
AY662080.1 | - | 7 - 8 | 892 | 82 | |
AY662082.1 | - | 7 - 8 | 892 | 82 | |
AY662083.1 | - | 7 - 8 | 892 | 82 | |
AY662084.1 | - | 7 - 8 | 892 | 82 | |
D38416.1 | - | 7 - 8 | 907 | 82 |