The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291046.1 | NP_001277975.1 | 3 - 4 | 537 | 74 |
NM_019837.2 | NP_062811.1 | 3 - 4 | 529 | 74 | |
GenBank mRNA | AF264064.1 | - | 3 - 4 | 528 | 74 |
BC016534.1 | - | 4 - 5 | 435 | 74 | |
BC046805.1 | - | 3 - 4 | 530 | 74 | |
BU919402.1 | - | 3 - 4 | 361 | 74 |