Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fank1
Entrez Gene ID
66930
Gene Name
fibronectin type 3 and ankyrin repeat domains 1
Gene Aliases
1700007B22Rik, AI850911
Chromosome Location
Chr.7: 133776856 - 133881536 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025850.2 NP_080126.1 1 - 2 80 57
GenBank mRNA AK005694.1 - 1 - 2 90 57
AK017071.1 - 1 - 2 104 57
AY819025.1 - 1 - 2 110 57
BC046278.1 - 1 - 2 80 57

View Full Product Details