The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025968.3 | NP_080244.1 | 2 - 3 | 164 | 70 |
GenBank mRNA | AK010888.1 | - | 2 - 3 | 199 | 70 |
AK011962.1 | - | 2 - 3 | 186 | 70 | |
AK035425.1 | - | 2 - 3 | 165 | 70 | |
AK134440.1 | - | 2 - 3 | 197 | 70 | |
AK166835.1 | - | 2 - 3 | 164 | 70 | |
BC014865.1 | - | 2 - 3 | 146 | 70 | |
CT010267.1 | - | 1 - 2 | 113 | 70 |