The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007802.4 | NP_031828.2 | 7 - 8 | 1019 | 73 |
GenBank mRNA | AK003425.1 | - | 7 - 8 | 944 | 73 |
AK132648.1 | - | 7 - 8 | 949 | 73 | |
BC046320.1 | - | 7 - 8 | 973 | 73 | |
X94444.1 | - | 6 - 7 | 891 | 73 |