Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4a14
Entrez Gene ID
13119
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 14
Gene Aliases
AI314743
Chromosome Location
Chr.4: 115486200 - 115496159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007822.2 NP_031848.1 9 - 10 1243 94
GenBank mRNA AK098088.1 - 9 - 10 1240 94
AK149392.1 - 9 - 10 1241 94
AK165452.1 - 9 - 10 1241 94
BC089609.1 - 9 - 10 1243 94

View Full Product Details