The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198949.1 | NP_001185878.1 | 4 - 5 | 1351 | 80 |
NM_009067.5 | NP_033093.2 | 4 - 5 | 1675 | 80 | |
GenBank mRNA | AK145387.1 | - | 4 - 5 | 1215 | 80 |
BC054329.1 | - | 4 - 5 | 1266 | 80 | |
BC067073.1 | - | 3 - 4 | 1110 | 80 | |
BC075732.1 | - | 4 - 5 | 1300 | 80 | |
BC076636.1 | - | 4 - 5 | 1267 | 80 | |
BC138546.1 | - | 3 - 4 | 1116 | 80 | |
BC145815.1 | - | 3 - 4 | 1116 | 80 | |
X80937.1 | - | 4 - 5 | 1252 | 80 |