Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfrp2
Entrez Gene ID
20319
Gene Name
secreted frizzled-related protein 2
Gene Aliases
AI851596, Sdf5
Chromosome Location
Chr.3: 83766148 - 83774314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009144.2 NP_033170.1 1 - 2 727 93
GenBank mRNA AF017989.1 - 1 - 2 759 93
AK138252.1 - 1 - 2 566 93
AK147091.1 - 1 - 2 724 93
BC014722.1 - 1 - 2 687 93
D50462.1 - 1 - 2 519 93
U88567.1 - 1 - 2 741 93

View Full Product Details