The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009180.3 | NP_033206.2 | 8 - 9 | 1063 | 108 |
GenBank mRNA | AK004613.1 | - | 8 - 9 | 1064 | 108 |
AK079216.1 | - | - | 1221 | 108 | |
AK136381.1 | - | - | 1066 | 108 | |
BC003276.1 | - | 8 - 9 | 1965 | 108 | |
BC010208.1 | - | - | 1015 | 108 | |
BC023076.1 | - | 2 - 3 | 1511 | 108 | |
EU937514.1 | - | 2 - 3 | 1510 | 108 | |
X93999.1 | - | 8 - 9 | 1027 | 108 |