The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011840.2 | NP_035970.1 | 21 - 22 | 1829 | 76 |
GenBank mRNA | AB019374.1 | - | 21 - 22 | 1830 | 76 |
AK159261.1 | - | 21 - 22 | 1879 | 76 | |
BC028260.1 | - | 20 - 21 | 1682 | 76 |