The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018881.3 | NP_061369.2 | 2 - 3 | 175 | 76 |
GenBank mRNA | AF184981.1 | - | 1 - 2 | 240 | 76 |
AK009224.1 | - | 2 - 3 | 210 | 76 | |
BC031415.1 | - | 2 - 3 | 175 | 76 |