Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo2
Entrez Gene ID
55990
Gene Name
flavin containing monooxygenase 2
Gene Aliases
2310008D08Rik, 2310042I22Rik, AW107733
Chromosome Location
Chr.1: 162874321 - 162898712 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018881.3 NP_061369.2 7 - 8 1229 76
GenBank mRNA AF184981.1 - 6 - 7 1294 76
AK129026.1 - 3 - 4 762 76
BC031415.1 - 7 - 8 1229 76

View Full Product Details