Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syap1
Entrez Gene ID
67043
Gene Name
synapse associated protein 1
Gene Aliases
2010110O17Rik, AW011796
Chromosome Location
Chr.X: 162856843 - 162888462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025932.2 NP_080208.2 4 - 5 653 82
GenBank mRNA AK008390.1 - 4 - 5 622 82
AK014893.1 - 4 - 5 639 82
AK051251.1 - 4 - 5 610 82
AK147055.1 - 4 - 5 589 82
AK154843.1 - 4 - 5 610 82
BC021373.1 - 4 - 5 583 82

View Full Product Details