Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syap1
Entrez Gene ID
67043
Gene Name
synapse associated protein 1
Gene Aliases
2010110O17Rik, AW011796
Chromosome Location
Chr.X: 162856843 - 162888462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025932.2 NP_080208.2 6 - 7 946 96
GenBank mRNA AK008390.1 - 6 - 7 915 96
AK014893.1 - 6 - 7 932 96
AK051251.1 - 6 - 7 903 96
AK147055.1 - 6 - 7 882 96
AK154843.1 - 6 - 7 903 96
BC021373.1 - 6 - 7 876 96

View Full Product Details