Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xlr3c
Entrez Gene ID
22446
Gene Name
X-linked lymphocyte-regulated 3C
Gene Aliases
Xlr3b
Chromosome Location
Chr.X: 73254538 - 73265571 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011727.2 NP_035857.2 3 - 4 247 89
GenBank mRNA L22977.1 - - 232 89
U02600.1 - 3 - 4 247 89

Gene Symbol
Xlr3a
Entrez Gene ID
22445
Gene Name
X-linked lymphocyte-regulated 3A
Gene Aliases
Xlr3
Chromosome Location
Chr.X: 73086293 - 73097095 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110784.1 NP_001104254.1 3 - 4 307 89
GenBank mRNA BC003812.1 - 3 - 4 253 89
BC093495.1 - - 230 89
BC107255.1 - - 216 89
BC107283.1 - - 217 89
BC127145.1 - - 217 89
BC127146.1 - - 218 89
BQ885410.1 - 3 - 4 247 89
DV662513.1 - 3 - 4 255 89
U02599.1 - 2 - 3 200 89

Gene Symbol
Xlr3b
Entrez Gene ID
574437
Gene Name
X-linked lymphocyte-regulated 3B
Gene Aliases
EG574437, Xlr3
Chromosome Location
Chr.X: 73192152 - 73202930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081643.1 NP_001075112.1 3 - 4 278 89
GenBank mRNA AK146398.1 - 3 - 4 232 89
BC038862.1 - 3 - 4 241 89
BC064787.1 - 3 - 4 235 89
CJ063846.1 - 3 - 4 279 89

View Full Product Details