Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk7
Entrez Gene ID
23993
Gene Name
kallikrein related-peptidase 7 (chymotryptic, stratum corneum)
Gene Aliases
Prss6, SCCE
Chromosome Location
Chr.7: 43811444 - 43816359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011872.2 NP_036002.1 4 - 5 547 115
GenBank mRNA AB008371.1 - 4 - 5 524 115
AF124299.1 - 4 - 5 549 115
AK029477.1 - 4 - 5 547 115
AK077406.1 - 4 - 5 547 115
BC027823.1 - 4 - 5 708 115

View Full Product Details