The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080793.5 | NP_542983.3 | 1 - 2 | 307 | 62 |
GenBank mRNA | AF448509.1 | - | 1 - 2 | 89 | 62 |
AK048924.1 | - | 1 - 2 | 144 | 62 | |
AK129428.1 | - | 1 - 2 | 96 | 62 | |
AK147316.1 | - | 1 - 2 | 363 | 62 | |
AK147413.1 | - | 1 - 2 | 133 | 62 | |
AK147422.1 | - | 1 - 2 | 308 | 62 | |
AK147667.1 | - | 1 - 2 | 342 | 62 | |
AK170161.1 | - | 1 - 2 | 151 | 62 | |
BC050190.1 | - | 1 - 2 | 131 | 62 |