Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rela
Entrez Gene ID
19697
Gene Name
v-rel reticuloendotheliosis viral oncogene homolog A (avian)
Gene Aliases
p65
Chromosome Location
Chr.19: 5637440 - 5648130 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009045.4 NP_033071.1 10 - 11 1374 64
GenBank mRNA AK089489.1 - 10 - 11 1119 64
AK154630.1 - 10 - 11 1119 64
AK155321.1 - 10 - 11 1383 64
AK155889.1 - 10 - 11 1116 64
AK157076.1 - 10 - 11 1106 64
AK158072.1 - 10 - 11 1384 64
BC003818.1 - 10 - 11 1133 64
BC094053.1 - 10 - 11 1374 64
DQ020177.1 - 8 - 9 943 64
M61909.1 - 10 - 11 1089 64

View Full Product Details