The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133737.2 | NP_598498.1 | 2 - 3 | 924 | 67 |
GenBank mRNA | AB035967.1 | - | 2 - 3 | 743 | 67 |
AK005632.1 | - | 2 - 3 | 293 | 67 | |
BC016072.1 | - | 2 - 3 | 778 | 67 |