Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc59
Entrez Gene ID
98238
Gene Name
leucine rich repeat containing 59
Gene Aliases
AA959742, C78668
Chromosome Location
Chr.11: 94629824 - 94645216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133807.1 NP_598568.1 1 - 2 289 78
GenBank mRNA AK053450.1 - 1 - 2 316 78
AK159374.1 - 1 - 2 345 78
AK159552.1 - 1 - 2 339 78
AK166576.1 - 1 - 2 338 78
AK167604.1 - 1 - 2 280 78
AK169250.1 - 1 - 2 264 78
AK169390.1 - 1 - 2 288 78
AK183165.1 - 1 - 2 75 78
BC006877.1 - 1 - 2 289 78
BC066172.1 - 1 - 2 283 78

View Full Product Details