Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Plekha1
- Entrez Gene ID
-
101476
- Gene Name
- pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 1
- Gene Aliases
-
AA960558, C920009D07Rik, TAPP1
- Chromosome Location
-
Chr.7: 130865729 - 130913308 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_133942.2 |
NP_598703.1 |
3 - 4 |
289 |
63 |
GenBank mRNA |
BC020017.1 |
- |
3 - 4 |
289 |
63 |
View Full Product Details