The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133948.5 | NP_598709.1 | 10 - 11 | 1290 | 89 |
GenBank mRNA | AF339082.1 | - | 10 - 11 | 1337 | 89 |
AJ308965.1 | - | 9 - 10 | 980 | 89 | |
BC043079.1 | - | 10 - 11 | 1237 | 89 |