Product Details
- Assay Reference Genome
Location
Chr.19:44833295 on build GRCm38
- Cytoband
- 19C3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 73322
- Gene Symbol
- 1700039E22Rik
- Gene Name
- RIKEN cDNA 1700039E22 gene
- Gene Aliases
- Location
- Chr.19:44828493-44835938 on build GRCm38
- Assay Gene Location
- Within Intron 1
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
1700039E22Rik |
NR_045315.1 |
|
|
|
AK006641.1 |
|
|
|
- Entrez Gene ID
- 18504
- Gene Symbol
- Pax2
- Gene Name
- paired box 2
- Gene Aliases
- Opdc, Pax-2
- Location
- Chr.19:44745695-44837871 on build GRCm38
- Assay Gene Location
- Overlaps Exon 8 - Intron 8
View Full Product Details