Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cinp
Entrez Gene ID
67236
Gene Name
cyclin-dependent kinase 2 interacting protein
Gene Aliases
1810047K05Rik, 2810452K22Rik
Chromosome Location
Chr.12: 110872610 - 110889172 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026048.4 NP_080324.1 3 - 4 382 69
GenBank mRNA AK004355.1 - 3 - 4 337 69
AK076126.1 - 3 - 4 352 69
AK160662.1 - 3 - 4 352 69
BC011066.1 - 3 - 4 352 69

View Full Product Details