The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252448.1 | NP_001239377.1 | 12 - 13 | 1319 | 93 |
NM_028126.3 | NP_082402.1 | 10 - 11 | 1208 | 93 | |
GenBank mRNA | AK011451.1 | - | 10 - 11 | 1154 | 93 |
AK138358.1 | - | 12 - 13 | 1320 | 93 | |
AY341872.1 | - | 10 - 11 | 1167 | 93 | |
BC052063.1 | - | - | 1215 | 93 | |
BC058517.1 | - | 10 - 11 | 1109 | 93 |